Indium antimonide ≥99.99% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-4920-25G 93-4920-5G
100203-684EA 720.43 USD
100203-684 100198-400
Indium antimonide ≥99.99% (In basis), PURATREM
Indium antimonide
Formula: InSb
MW: 236.58 g/mol
Melting Pt: 527 °C
Density: 5.78 g/cm³ (20 °C)
MDL Number: MFCD00016146
CAS Number: 1312-41-0
EINECS: 215-192-3
UN: 1549
ADR: 6.1,III
Merck Index: 13,04972

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR