Indium(III) iodide ≥99.999% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

93-4915-25G 93-4915-1G 93-4915-5G
100209-280EA 1106.83 USD
100209-280 100200-154 100200-156
Indium(III) iodide ≥99.999% (In basis), PURATREM
Indium(III) iodide
Formula: I₃In
MW: 495.53 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 209.85 °C
Density: 4.69 g/cm³ (20 °C)
MDL Number: MFCD00016151
CAS Number: 13510-35-5
EINECS: 236-839-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR