Indium(III) sulfide ≥99.99% (In basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Diindium trisulphide, Indium trisulphide
93-4911-5G
93-4911-25G
100198-398EA
198.15
USD
100198-398
100210-694
Indium(III) sulfide ≥99.99% (In basis), PURATREM
Indium(III) sulphide
Formula:
In₂S₃ MW: 325.84 g/mol Melting Pt: 1080 °C Density: 4.89 g/cm³ (25 °C) |
MDL Number:
MFCD00011061 CAS Number: 12030-24-9 EINECS: 234-742-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...