Indium(III) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Indium (III) tris(2,4-pentanedionato-O,O'), Indium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')indium (III)

93-4901-25G 93-4901-5G
100206-038EA 508.12 USD
100206-038 100202-160
Indium(III) acetylacetonate ≥98%
Indium(III) acetylacetonate
Formula: C₁₅H₂₁InO₆
MW: 412.15 g/mol
Melting Pt: 180…185 °C
MDL Number: MFCD00013494
CAS Number: 14405-45-9
EINECS: 238-378-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR