Cadmium oxide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cadmium (II) oxide

93-4835-1000G 93-4835-250G
100202-154EA 236.37 USD
100202-154 100199-114
Cadmium oxide ≥99%
Cadmium oxide
Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1559 °C (1013 hPa)
Melting Pt: ~ 1426 °C
Density: 8.15 g/cm³ (20 °C)
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,I
Merck Index: 13,01623

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR