Cadmium sulphide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cadmium (II) sulphide

93-4823-500G 93-4823-100G 93-4823-25G
100202-804EA 329.78 USD
100202-804 100205-604 100205-606
Cadmium sulphide ≥98%
Cadmium sulphide
Formula: CdS
Melting Pt: 1400 °C
Density: 4.82 g/cm³ (20 °C)
MDL Number: MFCD00010922
CAS Number: 1306-23-6
EINECS: 215-147-8
UN: 2570
ADR: 9,III
Merck Index: 12,01666

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR