Cadmium(II) sulfate hydrate ≥98% ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-4821-250G 93-4821-50G
100204-726EA 147.2 USD
100204-726 100197-868
Cadmium(II) sulfate hydrate ≥98% ACS
Cadmium(II) sulphate hydrate
Formula: CdO₄S·H₂O
MW: 226.49 g/mol
Density: 3.08 g/cm³ (20 °C)
MDL Number: MFCD00151655
CAS Number: 15244-35-6
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR