Cadmium oxide ≥99.99% (Cd basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cadmium (II) oxide

93-4817-250G 93-4817-1000G
100204-348EA 301.48 USD
100204-348 100197-238
Cadmium oxide ≥99.99% (Cd basis), PURATREM
Cadmium oxide
Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1559 °C (1013 hPa)
Melting Pt: 1430 °C
Density: 8.15 g/cm³ (20 °C)
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,III
Merck Index: 13,01623

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR