Cadmium(II) nitrate tetrahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-4815-500G 93-4815-100G
100205-600EA 181.17 USD
100205-600 100204-346
Cadmium(II) nitrate tetrahydrate ≥98%
Cadmium(II) nitrate tetrahydrate
Formula: CdN₂O₆·4H₂O
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: 59.4 °C
Density: 2.45
MDL Number: MFCD00149626
CAS Number: 10022-68-1
EINECS: 233-710-6
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR