Cadmium(II) iodide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-4812-25G 93-4812-100G
100203-488EA 76.43 USD
100203-488 100209-436
Cadmium(II) iodide ≥99%
Cadmium(II) iodide
Formula: CdI₂
Boiling Pt: 787 °C (1013 hPa)
Melting Pt: 387 °C
Density: 5.67 g/cm³ (20 °C)
MDL Number: MFCD00010920
CAS Number: 7790-80-9
EINECS: 232-223-6
UN: 2570
ADR: 6.1,III
Merck Index: 12,01657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR