Cadmium acetate dihydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate
93-4801-250G
93-4801-1000G
100210-884EA
257.6
USD
100210-884
100197-236
Cadmium acetate dihydrate ≥98%
Cadmium acetate dihydrate
Formula:
(H₃CCOO)₂Cd·2H₂O MW: 266.53 g/mol Boiling Pt: 105 - 107 °C (4 -rr) Melting Pt: 256 °C Density: 2.341 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150020 CAS Number: 5743-04-4 EINECS: 208-853-2 UN: 2570 ADR: 6.1,III Merck Index: 13,01614 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...