Silver ≥99.9%, flakes

Supplier: STREM CHEMICALS INC MS

93-4760-5G 93-4760-25G
100196-688EA 56.62 USD
100196-688 100198-392
Silver ≥99.9%, flakes
Silver
Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960.5 °C
Density: 10.49
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR