Silver ≥99.9%, Powder
Supplier: STREM CHEMICALS INC MS
93-4758-5G
93-4758-25G
100207-084EA
42.46
USD
100207-084
100203-484
Silver ≥99.9%, Powder
Silver
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960.5 °C Density: 10.49 Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...