Silver(I) oxide ≥99% (99.99%-Ag), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4743-50G
93-4743-10G
100211-264EA
308.55
USD
100211-264
100202-144
Silver(I) oxide ≥99% (99.99%-Ag), PURATREM
Silver(I) oxide
Formula:
Ag₂O Melting Pt: 300 °C (dec.) Density: 7.143 Storage Temperature: Refrigerator |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 EINECS: 243-957-1 UN: 1479 ADR: 5.1,II Merck Index: 12,08665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...