Silver(II) oxide ≥99.9% (Ag basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4742-10G
93-4742-50G
100211-262EA
100.49
USD
100211-262
100202-454
Silver(II) oxide ≥99.9% (Ag basis)
Silver(II) oxide
Formula:
AgO MW: 123.87 g/mol Melting Pt: > 100 °C (dec.) Density: 7.44 g/cm³ (20 °C) |
MDL Number:
MFCD00044285 CAS Number: 1301-96-8 EINECS: 215-098-2 UN: 1479 ADR: 5.1,II Merck Index: 12,08666 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...