Silver sulfate ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4731-10G
93-4731-50G
100200-580EA
121.72
USD
100200-580
100210-692
Silver sulfate ≥98% ACS
Silver sulphate
Formula:
Ag₂SO₄ Boiling Pt: 1085 °C (dec.) Melting Pt: 652 °C Density: 5.45 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003407 CAS Number: 10294-26-5 EINECS: 233-653-7 Merck Index: 13,08603 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...