Silver perchlorate, anhydrous ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

93-4722-5G 93-4722-25G
100202-140EA 90.58 USD
100202-140 100202-138
Silver perchlorate, anhydrous ≥97%
Silver perchlorate
Formula: AgClO₄
MW: 207.32 g/mol
Melting Pt: 486 (dec.)
Density: 2.806
MDL Number: MFCD00003400
CAS Number: 7783-93-9
EINECS: 232-035-4
UN: 1481
ADR: 5.1 II
Merck Index: 12,08667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR