Silver nitrite ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Argentous nitrite

93-4719-10G 93-4719-50G
100205-590EA 108.98 USD
100205-590 100208-724
Silver nitrite ≥99%
Silver nitrite
Formula: AgNO₂
MW: 153.87 g/mol
Melting Pt: 140 °C
Density: 4.45 g/cm³ (20 °C)
MDL Number: MFCD00003413
CAS Number: 7783-99-5
EINECS: 232-041-7
UN: 2627
ADR: 5.1,II
Merck Index: 12,08663

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR