Silver chloride ≥99.9% (Ag basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-4707-10G 93-4707-50G
100196-676EA 84.92 USD
100196-676 100210-418
Silver chloride ≥99.9% (Ag basis)
Silver chloride
Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1550 °C
Melting Pt: 455 °C
Density: 5.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
Merck Index: 13,08582

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR