Sodium tetrachloropalladate(II) trihydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-4613-2G 93-4613-10G
100207-078EA 257.6 USD
100207-078 100207-076
Sodium tetrachloropalladate(II) trihydrate ≥99%
Sodium tetrachloropalladate(II) trihydrate
Formula: Na₂PdCl₄·3H₂O
MW: 348.22 g/mol
MDL Number: MFCD00149171
CAS Number: 13820-53-6
EINECS: 237-502-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR