Sodium hexachloropalladate(IV) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Disodium hexachloropalladate
93-4612-1G
93-4612-5G
100196-666EA
217.97
USD
100196-666
100208-182
Sodium hexachloropalladate(IV) ≥98%
Sodium hexachloropalladate(IV)
Formula:
Cl₆Na₂Pd MW: 365.12 g/mol |
MDL Number:
MFCD00014235 CAS Number: 53823-60-2 EINECS: 258-809-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...