Diammonium tetrachloropalladate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4602-1G
93-4602-5G
100200-576EA
113.23
USD
100200-576
100205-582
Diammonium tetrachloropalladate ≥99%
Diammonium tetrachloropalladate
Formula:
H₈Cl₄N₂Pd Melting Pt: Dec. Density: 2.17 g/cm³ (20 °C) |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 EINECS: 237-498-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...