Ammonium hexachloropalladate(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Diammonium hexachloropalladate

93-4601-1G 93-4601-5G
100200-574EA 108.98 USD
100200-574 100208-564
Ammonium hexachloropalladate(IV) ≥99%
Ammonium hexachloropalladate(IV)
Formula: H₈Cl₆N₂Pd
MW: 355.21 g/mol
Melting Pt: Dec.
Density: 2.418 g/cm³ (20 °C)
MDL Number: MFCD00015959
CAS Number: 19168-23-1
EINECS: 242-854-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR