Molybdenum ≥99.97%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4259-1000MT
93-4259-250MT
100207-220EA
668.06
USD
100207-220
100202-136
Molybdenum ≥99.97%, wire
Molybdenum
Formula:
Mo Boiling Pt: 5560 °C Melting Pt: 2610 °C Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...