Molybdenum ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-4257-500G 93-4257-100G
100202-798EA 343.94 USD
100202-798 100207-064
Molybdenum ≥99.9%, powder
Molybdenum
Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.2
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4 II
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR