Molybdenum ≥99.95%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4256-500G
93-4256-100G
100203-472EA
489.72
USD
100203-472
100204-330
Molybdenum ≥99.95%, powder
Molybdenum
Formula:
Mo MW: 95.94 g/mol Boiling Pt: 3617 Melting Pt: 2622 Density: 10.2 Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 UN: 3089 ADR: 4 II Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...