Molybdenum dioxide ≥99%

Supplier: STREM CHEMICALS INC MS

93-4246-50G 93-4246-10G
100207-062EA 108.98 USD
100207-062 100210-410
Molybdenum dioxide ≥99%
Molybdenum dioxide
Formula: MoO₂
MW: 127.94 g/mol
Boiling Pt: 1207 - 1307 °C (1013 hPa)
Melting Pt: 770 - 795 °C
Density: 6.47 g/cm³ (25 °C)
MDL Number: MFCD00014223
CAS Number: 18868-43-4
EINECS: 242-637-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR