Molybdenum diselenide ≥99.9% (Mo basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4211-25G
93-4211-5G
100210-406EA
475.57
USD
100210-406
100210-408
Molybdenum diselenide ≥99.9% (Mo basis)
Molybdenum diselenide
Formula:
MoSe₂ MW: 253.86 Melting Pt: 1200 Density: 6.0 |
MDL Number:
MFCD00014216 CAS Number: 12058-18-3 EINECS: 235-027-9 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...