Niobium(V) oxide 99,9 Nb basis

Supplier: STREM CHEMICALS INC MS

Synonyms: Diniobium pentaoxide, Niobium pentoxide

93-4109-250G 93-4109-50G
100206-032EA 294.4 USD
100206-032 100202-124
Niobium(V) oxide 99,9 Nb basis
Niobium(V) oxide
Formula: Nb₂O₅
MW: 265.81 g/mol
Melting Pt: 1496
Density: 4.47
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR