Zirconium ≥99.5%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-4047-100G 93-4047-500G
100205-570EA 182.58 USD
100205-570 100204-722
Zirconium ≥99.5%, powder
Zirconium
Formula: Zr
MW: 91.22 g/mol
Boiling Pt: 3577 °C
Melting Pt: 1852 °C
Density: 6.506
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9
UN: 2008
ADR: 4.2 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR