Zirconium tetrachloride ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Zirconium (IV) chloride
93-4045-50G
93-4045-250G
100199-100EA
42.46
USD
100199-100
100208-562
Zirconium tetrachloride ≥99.5%
Zirconium tetrachloride
Formula:
Cl₄Zr MW: 233.03 g/mol Boiling Pt: 3577 (subl.) Melting Pt: 437 Density: 2.803 Storage Temperature: Ambient |
MDL Number:
MFCD00011306 CAS Number: 10026-11-6 EINECS: 233-058-2 UN: 2503 ADR: 8 III Merck Index: 12,10307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...