Zirconyl nitrate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate

93-4042-50G 93-4042-250G
100196-650EA 134.46 USD
100196-650 100201-280
Zirconyl nitrate hydrate
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
MW: 249.25 g/mol
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 2728
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR