Zirconium nitride 98

Supplier: STREM CHEMICALS INC MS

93-4034-50G 93-4034-10G
100202-796EA 315.63 USD
100202-796 100205-568
Zirconium nitride 98
Zirconium nitride
Formula: NZr
MW: 105.23
Melting Pt: 2980
Density: 7.09
MDL Number: MFCD00049642
CAS Number: 25658-42-8
EINECS: 247-166-2
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR