Zirconium tetraiodide ≥99.5% (Zr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-4025-5G
93-4025-1G
100200-340EA
256.18
USD
100200-340
100200-566
Zirconium tetraiodide ≥99.5% (Zr basis)
Zirconium tetraiodide
Formula:
I₄Zr MW: 598.84 Melting Pt: 499 |
MDL Number:
MFCD00049641 CAS Number: 13986-26-0 EINECS: 237-780-9 UN: 3260 ADR: 8 III Merck Index: 12,10311 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...