Zirconium tetrafluoride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zirconium (IV) fluoride

93-4023-100G 93-4023-25G
100203-466EA 227.88 USD
100203-466 100202-122
Zirconium tetrafluoride ≥98%
Zirconium tetrafluoride
Formula: F₄Zr
MW: 167.22 g/mol
Melting Pt: ~600 °C (subl.)
Density: 4.43
MDL Number: MFCD00011307
CAS Number: 7783-64-4
EINECS: 232-018-1
UN: 3260
ADR: 8 II
Merck Index: 12,10308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR