Yttrium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

93-3944-25X25MM 93-3944-50X50MM
100210-392EA 246.28 USD
100210-392 100207-054
Yttrium ≥99.9%, foil
Yttrium
Formula: Y
Boiling Pt: 2927 °C (1013 hPa)
Melting Pt: 1525 °C
Density: 4.472 g/cm³ (20 °C)
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR