Yttrium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS
Warning

93-3942-10G 93-3942-50G
100207-050EA 137.29 USD
100207-050 100200-140
Yttrium ≥99.9%, ingot
Yttrium
Formula: Y
Boiling Pt: 3380 °C
Melting Pt: 1522 °C
Density: 4.472 g/cm³ (20 °C)
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR