Yttrium ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3940-25G
93-3940-5G
100196-648EA
462.83
USD
100196-648
100197-842
Yttrium ≥99.9%, powder
Yttrium
Formula:
Y Boiling Pt: 3380 °C Melting Pt: 1522 °C Density: 4,47 |
MDL Number:
MFCD00011468 CAS Number: 7440-65-5 EINECS: 231-174-8 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...