Yttrium (III) nitrate hexahydrate ≥99.9% (Y basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Yttrium trinitrate hexahydrate

93-3937-250G 93-3937-50G
100203-460EA 329.78 USD
100203-460 100203-462
Yttrium (III) nitrate hexahydrate ≥99.9% (Y basis)
Yttrium (III) nitrate hexahydrate
Formula: N₃O₉Y·6H₂O
MW: 383.01 g/mol
Melting Pt: 51.8 °C
Density: 2.682 g/cm³ (20 °C)
MDL Number: MFCD00149940
CAS Number: 13494-98-9
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR