Yttrium(III) fluoride ≥99.9% (Y basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3936-10G
93-3936-50G
100205-558EA
43.88
USD
100205-558
100208-168
Yttrium(III) fluoride ≥99.9% (Y basis)
Yttrium(III) fluoride
Formula:
F₃Y Boiling Pt: 2227 °C (1013 hPa) Melting Pt: 1152 °C Density: 4.01 |
MDL Number:
MFCD00049714 CAS Number: 13709-49-4 EINECS: 237-257-5 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...