Yttrium(III) chloride, anhydrous ≥99.9% (Y basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-3935-10G 93-3935-50G
100199-094EA 82.09 USD
100199-094 100207-048
Yttrium(III) chloride, anhydrous ≥99.9% (Y basis)
Yttrium(III) chloride
Formula: Cl₃Y
MW: 195.26
Boiling Pt: 1488
Melting Pt: 703
Density: 2.67
Storage Temperature: Ambient
MDL Number: MFCD00011470
CAS Number: 10361-92-9
EINECS: 233-801-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR