Diyttrium trioxide ≥99.99% (Y basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Yttrium oxide, Yttrium (III) oxide
93-3910-50G
93-3910-250G
100197-840EA
63.69
USD
100197-840
100205-556
Diyttrium trioxide ≥99.99% (Y basis), PURATREM
Diyttrium trioxide
Formula:
Y₂O₃ MW: 225.81 g/mol Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 2410 °C Density: 5.01 g/cm³ (20 °C) |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...