Yttrium(III) chloride hexahydrate ≥99.9% (Y basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Yttrium trichloride hexahydrate
93-3903-100G
93-3903-25G
100207-840EA
172.68
USD
100207-840
100210-386
Yttrium(III) chloride hexahydrate ≥99.9% (Y basis)
Yttrium(III) chloride hexahydrate
Formula:
YCl₃·6H₂O MW: 303.36 Boiling Pt: 1507 Melting Pt: 153 Density: 2.18 |
MDL Number:
MFCD00149941 CAS Number: 10025-94-2 EINECS: 233-801-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...