Strontium acetylacetonate hydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(2,4-pentanedionato-O,O')strontium hydrate, Strontium bis(2,4-pentanedionato-O,O') hydrate, Strontium 2,4-pentanedionate hydrate

93-3847-100G 93-3847-25G
100202-110EA 703.45 USD
100202-110 100204-720
Strontium acetylacetonate hydrate ≥99%
Strontium acetylacetonate hydrate
Formula: C₁₀H₁₄O₄Sr·H₂O
MW: 303.85
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00013502
CAS Number: 12193-47-4
EINECS: 235-366-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR