Strontium ≥99%, pieces

Supplier: STREM CHEMICALS INC MS
Danger

93-3831-100G 93-3831-25G
100208-716EA 549.17 USD
100208-716 100202-108
Strontium ≥99%, pieces
Strontium
Formula: Sr
Boiling Pt: 1151 °C (1013 hPa)
Melting Pt: 570 - 660 °C
Density: 2.38 g/cm³ (770 °C)
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3,II
Merck Index: 12,08994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR