Strontium titanate ≥99%

Supplier: STREM CHEMICALS INC MS

93-3823-1000G 93-3823-250G
100205-554EA 225.05 USD
100205-554 100196-642
Strontium titanate ≥99%
Strontium titanate
Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5,11 g/cm³ (20 °C)
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR