Strontium sulphide ≥99.9% (Sr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3821-50G
93-3821-10G
100199-280EA
656.74
USD
100199-280
100204-718
Strontium sulphide ≥99.9% (Sr basis)
Strontium sulphide
Formula:
SSr Melting Pt: > 2000 °C Density: 3.7 g/cm³ (20 °C) |
MDL Number:
MFCD00049560 CAS Number: 1314-96-1 EINECS: 215-249-2 UN: 3134 ADR: 4.3,III Merck Index: 12,09014 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...