Strontium perchlorate hydrate ≥99.9% (Sr basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-3817-100G 93-3817-500G
100209-106EA 148.62 USD
100209-106 100200-754
Strontium perchlorate hydrate ≥99.9% (Sr basis)
Strontium perchlorate hydrate
Formula: Cl₂O₈Sr·2H₂O
MW: 286.52 g/mol (anhy)
Melting Pt: < 100 °C
MDL Number: MFCD09039173
CAS Number: 13450-97-0
EINECS: 236-614-2
UN: 1508
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR