Strontium chloride, anhydrous ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

93-3806-1000G 93-3806-250G
100200-136EA 281.66 USD
100200-136 100200-556
Strontium chloride, anhydrous ≥95%
Strontium chloride
Formula: Cl₂Sr
Boiling Pt: 1250 °C (1013 hPa)
Melting Pt: 868 °C
Density: 1.96 g/cm³ (20 °C)
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR