Strontium bromide, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-3802-100G 93-3802-25G
100199-084EA 230.71 USD
100199-084 100210-374
Strontium bromide, anhydrous ≥99%
Strontium bromide
Formula: Br₂Sr
Boiling Pt: 2146 °C (1013 hPa)
Melting Pt: 643 °C
Density: 4.216
MDL Number: MFCD00049548
CAS Number: 10476-81-0
EINECS: 233-969-5
Merck Index: 13,08917

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR